Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AF4 Monoclonal Antibody (C2.1.1), QED Bioscience™
SDP

Catalog No. 502277989
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7989 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7989 Supplier Leinco Technologies Inc Supplier No. 34042100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

AF4 Monoclonal antibody specifically detects AF4 in Human samples. It is validated for Western Blot

Specifications

Antigen AF4
Applications Western Blot
Classification Monoclonal
Clone C2.1.1
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene AFF1
Gene Accession No. P51825
Gene Alias 9630032B01Rik; AF4; AF4/FMR2 family member 1; AF4/FMR2 family, member 1; AFF1; ALL1-fused gene from chromosome 4 protein; AW319193; FEL; homolog of human MLLT2 unidentified; MLLT2; Mllt2h; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 2; myeloid/lymphoid or mixed-lineage leukemia translocated to 2 homolog; PBM1; pre-B-cell monocytic leukemia partner 1; protein AF-4; Protein FEL; Proto-oncogene AF4; Rob; robotic; translocated to, 2
Host Species Mouse
Immunogen Maltose-binding fusion protein containing AF4 amino acids 782-979 (Accession no. NP_005926).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4299
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.