Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AF4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562368
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2368 Supplier Bethyl Laboratories, Inc Supplier No. A302344A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Proto-oncogene AF-4 (or FEL) is a product of a chromosomal aberration of the human gene AFF1, which is associated with acute leukemias. The fusion of AF-4 on chromosome band 4q21 with the mixed lineage leukemia (MLL or HRX) gene on 11q23 results in a MLL-AF-4 chimeric transcription factor in which AF-4 contributes transcriptional effector properties and requires cell-specific accessory factors. MLL is involved in several chromosomal translocations associated with acute myeloid and lymphoid leukemia. The MLL-AF-4 fusion protein is expressed in all normal hematopoietic cells.

Specifications

Antigen AF4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene AFF1
Gene Accession No. P51825
Gene Alias MIXED LINEAGE LEUKEMIA, TRANSLOCATED TO, 2, ALL1 FUSED GENE FROM CHROMOSOME 4, AF4, FEL
Gene Symbols AFF1
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4299
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.