Learn More
AF4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302345A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Proto-oncogene AF-4 (or FEL) is a product of a chromosomal aberration of the human gene AFF1, which is associated with acute leukemias. The fusion of AF-4 on chromosome band 4q21 with the mixed lineage leukemia (MLL or HRX) gene on 11q23 results in a MLL-AF-4 chimeric transcription factor in which AF-4 contributes transcriptional effector properties and requires cell-specific accessory factors. MLL is involved in several chromosomal translocations associated with acute myeloid and lymphoid leukemia. The MLL-AF-4 fusion protein is expressed in all normal hematopoietic cells.Specifications
AF4 | |
Polyclonal | |
Unconjugated | |
AFF1 | |
MIXED LINEAGE LEUKEMIA, TRANSLOCATED TO, 2, ALL1 FUSED GENE FROM CHROMOSOME 4, AF4, FEL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4299 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P51825 | |
AFF1 | |
Between 175 and 225 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.