Learn More
AF6 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302199A

Description
The recommended shelf life for this product is 1 year from date of receipt.
AF-6, also known as afadin, is expressed in a variety of cell types. At least 5 isoforms of this protein are produced by alternative splicing and the ′canonical′ sequence is designated as l-afadin. Chromosomal aberration of the MLLT4 gene and tight junctions and Notch signaling pathways cross-communication. This happens through physical interaction with Ras, Notch and Dishevelled, a key Wingless effector. AF-6 also controls integrin-mediated cell adhesion. This is done at least partially by regulating Rap1 activation through recruitment of both SPA-1 and Rap1GTP. These results demonstrate that AF-6 is an important molecule mediating cell-cell adhesion but is also capable of modulating several key signaling pathways.Specifications
AF6 | |
Polyclonal | |
Unconjugated | |
AFDN | |
AF6, AF-6, ALL1-fused gene from chromosome 6 protein, MLL-AF6, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog); translocated to, 4, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4, protein AF-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4301 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P55196 | |
AFDN | |
Between 1475 and 1525 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.