Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AF9 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001565AF750
Description
AF9 Polyclonal antibody specifically detects AF9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
AF9 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AF-9, AF9myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 3, ALL1-fused gene from chromosome 9 protein, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 3, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 3, Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein, protein AF-9, YEATS domain-containing protein 3, YEATS3FLJ2035 | |
The immunogen recognized by this antibody maps to a region between residues 250 and 300 of human ALL1 Fused Gene from Chromosome 9 Protein using the numbering given in entry NP_004520.1 (GeneID 4300). | |
0.1 mL | |
Cancer | |
4300 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction