Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AF9 Antibody (HL1644) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325354
Description
AF9 Monoclonal antibody specifically detects AF9 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
AF9 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
HL1644 | |
Western Blot 1:500-1:3000, Immunocytochemistry/ Immunofluorescence | |
AF-9, AF9myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog),translocated to, 3, ALL1-fused gene from chromosome 9 protein, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog), translocatedto, 3, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila),translocated to, 3, Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein, protein AF-9, YEATS domain-containing protein 3, YEATS3FLJ2035 | |
Recombinant protein encompassing a sequence within the center region of human AF9. The exact sequence is proprietary. | |
100 μL | |
Cancer | |
4300 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction