Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AF9 (MLLT3) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE2597100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2597100 100 μg
ABE259725UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE2597100 Supplier MilliporeSigma Supplier No. ABE2597100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AF9 (MLLT3) Polyclonal antibody specifically detects AF9 (MLLT3) in Human, Mouse samples. It is validated for ChIP-Seq, Immunocytochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AF9 (MLLT3)
Applications ChIP sequencing (ChIP-seq), Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human AF9 (MLLT3) protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.