Learn More
Invitrogen™ AF9 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, HeLa, HepG2, THP-1, MLLT3 shRNA-transfected 293T, SEM-K2. Predicted reactivity: Mouse (98%), Rat (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | AF9 |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.46 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | MLLT3 |
| Gene Accession No. | A2AM29, P42568 |
| Gene Alias | 2210011H10Rik; 2610012I03Rik; 3830408D16Rik; Af9; Af-9; AF-9 protein; ALL1-fused gene from chromosome 9 protein; D4Ertd321e; MLLT3; MLLT3, super elongation complex subunit; myeloid/lymphoid or mixed lineage-leukemia translocation to 3 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein; Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein homolog; myeloid/lymphoid or mixed-lineage leukemia, translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia,translocated to, 3 (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia; translocated to 3; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 3; protein AF-9; translocated to, 3; YEATS domain-containing protein 3; YEATS3 |
| Gene Symbols | MLLT3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.