Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581972 Supplier Invitrogen™ Supplier No. PA581972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSKSDSEQPS PASSSSSSSS SFTPSQTRQQ GPLRSIMKDL HSDDNEEESD EVEDNDNDSE MERPVNRGGS RSRRVSLSDG SDSESSSASS PLHHEPPPPL LKTNNNQILE VKSPIKQSKS DKQIKNGECD KAYLDELVEL HRRLMTLR Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

The human AF9 gene is one of the most common fusion partner genes with the ALL1 gene at 11q23 (also called MLL), resulting in the t(9;11)(p22;q23). The AF9 gene is more than 100 kb, and 2 patient breakpoint cluster regions (BCRs) have been identified; BCR1 is within intron 4, previously called site A, whereas BCR2 or site B spans introns 7 and 8. Several different structural elements have been identified in AF9, including a colocalizing in vivo DNA topo II cleavage site and an in vitro DNase I hypersensitive (DNase 1 HS) site in intron 7 in BCR2. Reversibility experiments demonstrated a religation of the topo II cleavage sites. In addition, 2 scaffold associated regions (SARs) are located centromeric to the topo II and DNase I HS cleavage sites and border breakpoint regions in 2 leukemic cells lines: SAR1 is located in intron 4, whereas SAR2 encompasses parts of exons 5-7. The patient breakpoint regions of AF9 share the same structural elements as the MLL BCR. A DNA breakage and repair model for nonhomologous recombination between MLL and its partner genes, particularly AF9, has been proposed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AF9
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MLLT3
Gene Accession No. P42568
Gene Alias 2210011H10Rik; 2610012I03Rik; 3830408D16Rik; Af9; Af-9; AF-9 protein; ALL1-fused gene from chromosome 9 protein; D4Ertd321e; MLLT3; MLLT3, super elongation complex subunit; myeloid/lymphoid or mixed lineage-leukemia translocation to 3 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein; Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein homolog; myeloid/lymphoid or mixed-lineage leukemia, translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia,translocated to, 3 (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia; translocated to 3; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 3; protein AF-9; translocated to, 3; YEATS domain-containing protein 3; YEATS3
Gene Symbols MLLT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MLLT3. Recombinant protein control fragment (Product #RP-101898).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4300
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.