Learn More
AF9 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300595A
Description
The recommended shelf life for this product is 1 year from date of receipt.
The human AF9 gene is one of the most common fusion partner genes with the ALL1 gene at 11q23 (also called MLL), resulting in the t(9;11)(p22;q23). The AF9 gene is more than 100 kb, and 2 patient breakpoint cluster regions (BCRs) have been identified; BCR1 is within intron 4, previously called site A, whereas BCR2 or site B spans introns 7 and 8. Several different structural elements have been identified in AF9, including a colocalizing in vivo DNA topo II cleavage site and an in vitro DNase I hypersensitive (DNase 1 HS) site in intron 7 in BCR2. Reversibility experiments demonstrated a religation of the topo II cleavage sites. In addition, 2 scaffold associated regions (SARs) are located centromeric to the topo II and DNase I HS cleavage sites and border breakpoint regions in 2 leukemic cells lines: SAR1 is located in intron 4, whereas SAR2 encompasses parts of exons 5-7. The patient breakpoint regions of AF9 share the same structural elements as the MLL BCR. A DNA breakage and repair model for nonhomologous recombination between MLL and its partner genes, particularly AF9, has been proposed.Specifications
| AF9 | |
| Polyclonal | |
| Unconjugated | |
| MLLT3 | |
| AF-9 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4300, 70122 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| A2AM29, P42568 | |
| Mllt3 | |
| Between 200 and 250 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.