Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Afadin/AF-6 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP122996AF488
Description
Afadin/AF-6 Polyclonal antibody specifically detects Afadin/AF-6 in Human, Mouse samples. It is validated for Western BlotSpecifications
Afadin/AF-6 | |
Polyclonal | |
Western Blot | |
AF64, afadin, FLJ34371, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 4 | |
The immunogen recognized by this antibody maps to a region between residue 1475 and 1525 of human AF6 (myeloid/lymphoid or mixed-lineage leukemia translocated to, 4 ) using the numbering given in entry BAA32484.1 (GeneID 4301). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4301 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction