Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ l-Afadin Polyclonal Antibody

Catalog No. PIPA125075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA125075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA125075 Supplier Invitrogen™ Supplier No. PA125075

Rabbit Polyclonal Antibody

Recommended positive controls: MDCK, HepG2, mouse liver, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

AFDN belongs to an adhesion system, probably together with the E-cadherin-catenin system, which plays a role in the organization of homotypic, interneuronal and heterotypic cell-cell adherens junctions (AJs). Nectin- and actin-filament-binding protein that connects nectin to the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen l-Afadin
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 15mM sodium azide; pH 7.4
Gene AFDN
Gene Accession No. O35889, P55196, Q9QZQ1
Gene Alias 5033403D15Rik; AF6; Af-6; afadin; Afadin adherens junction formation factor; afadin, adherens junction formation factor; AFDN; ALL1-fused gene from chromosome 6 protein; Gm314; I-afadin; MLL-AF6; MLLT4; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; protein AF-6; S-afadin
Gene Symbols AFDN
Host Species Rabbit
Immunogen Synthetic peptide sequence corresponding to amino acid residues 1814-1829 of rat afadin with N-terminal cysteine conjugated to maleimide-activated KLH.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17356, 26955, 4301, 431695
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.