Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Afamin Antibody (120), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290199MFV450
Description
Afamin Monoclonal antibody specifically detects Afamin in Human samples. It is validated for ELISASpecifications
Afamin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
173 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
120 | |
ELISA | |
afamin, ALB2alpha-Alb, ALBAalpha-albumin, ALF, Alpha-Alb, Alpha-albumin, MGC125338, MGC125339 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Afamin (Uniprot#: P43652; Met1-Asn599). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction