Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Afamin Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA551041
Description
This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.
Specifications
Afamin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
P36953 | |
AFM | |
Recombinant Rat AFM protein, Leu22-Tyr201 (Accession #P36953). | |
200 μL | |
Primary | |
Rat | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
C1 | |
Unconjugated | |
AFM | |
Afamin; AFM; ALB2; ALBA; Alf; alpha albumin; alpha-Alb; Alpha-albumin; MGC125338; MGC125339 | |
Mouse | |
Protein A/G | |
RUO | |
282708 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction