Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Afamin Monoclonal Antibody (C12), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA551043
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551043 200 μL
FEMA551044 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551043 Supplier Invitrogen™ Supplier No. MA551043
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Afamin
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene AFM
Gene Accession No. P36953
Gene Alias Afamin; AFM; ALB2; ALBA; Alf; alpha albumin; alpha-Alb; Alpha-albumin; MGC125338; MGC125339
Gene Symbols AFM
Host Species Mouse
Immunogen Recombinant Rat AFM protein, Leu22-Tyr201 (Accession #P36953).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282708
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.