Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Afamin Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | Afamin |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5110823
|
Invitrogen™
PA5110823 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111355
|
Invitrogen™
PA5111355 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: EMEKLVKDMV EYKDRCMADK TLPECSKLPN NVLQEKICAM EGLPQKHNFS HCCSKVDAQR RLCFFYNKKS DVGFLPPFPT LDPEEKCQAY ESNRESLLNH FLYEVARRNP FVFAPTLLTV AVHFEEVAKS CCEEQNKVNC LQTRAIPV.
This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.Specifications
Afamin | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P43652 | |
173 | |
Recombinant Protein corresonding to Human Afamin. Recombinant protein control fragment (Product #RP-102380). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
AFM | |
Afamin; AFM; ALB2; ALBA; Alf; alpha albumin; alpha-Alb; Alpha-albumin; MGC125338; MGC125339 | |
AFM | |
Primary | |
Antigen Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title