Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AFAP Antibody (16F50), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222004IR
Description
AFAP Monoclonal specifically detects AFAP in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
AFAP | |
Monoclonal | |
DyLight 755 | |
Q12884 | |
AFAP1 | |
Human recombinant AFAP | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA | |
16F50 | |
Flow Cytometry, ELISA | |
actin filament associated protein 1, FLJ56849 | |
Mouse | |
Protein G purified | |
RUO | |
60312 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction