Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AFAP Antibody (OTI2G9), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272120UV
Description
AFAP Monoclonal specifically detects AFAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
AFAP | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AFAP1 | |
Recombinant protein fragment corresponding to amino acids 100-386 of human AFAP (NP_940997) produced in E.coli. | |
0.1 mL | |
60312.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2G9 | |
Western Blot, Immunohistochemistry | |
actin filament associated protein 1, FLJ56849 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction