Learn More
Invitrogen™ AFAP Monoclonal Antibody (OTI2G9)

Description
AFAP Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | AFAP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Afap1 |
| Gene Accession No. | Q8N556 |
| Gene Alias | 110 kDa actin filament-associated protein; 2600003E23Rik; 9630044L16Rik; actin filament associated protein 1; actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP; Afap1; AFAP110; AFAP-110; AI848729; FLJ56849; im:7148276; Kiaa3018; mKIAA3018 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.