Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AFAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544796 Supplier Invitrogen™ Supplier No. PA544796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QVKRKKSSKS EAKGTVSKVT GKKITKIISL GKKKPSTDEQ TSSAEEDVPT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFAP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Afap1
Gene Accession No. Q8N556
Gene Alias 110 kDa actin filament-associated protein; 2600003E23Rik; 9630044L16Rik; actin filament associated protein 1; actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP; Afap1; AFAP110; AFAP-110; AI848729; FLJ56849; im:7148276; Kiaa3018; mKIAA3018
Gene Symbols Afap1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human AFAP1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60312
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.