Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AFAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553356
Description
Immunogen sequence: PEALHYDYID VEMSASVIQT AKQTFCFMNR RVISANPYLG GTSNGYAHPS GTALHYDDVP CINGSLKGKK PPVASNGVTG KGKTLSSQPK KADPAAVVKR TGSNAAQYKY GKNRVEADAK R Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.
The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
AFAP | |
Polyclonal | |
Unconjugated | |
Afap1 | |
110 kDa actin filament-associated protein; 2600003E23Rik; 9630044L16Rik; actin filament associated protein 1; actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP; Afap1; AFAP110; AFAP-110; AI848729; FLJ56849; im:7148276; Kiaa3018; mKIAA3018 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60312 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N556 | |
Afap1 | |
Recombinant protein corresponding to Human AFAP. Recombinant protein control fragment (Product #RP-90783). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction