Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AFAP1L2 Antibody (2H6D1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | AFAP1L2 |
---|---|
Clone | 2H6D1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61836
![]() |
Novus Biologicals
NBP261836 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
AFAP1L2 Monoclonal specifically detects AFAP1L2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
AFAP1L2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
84632 | |
Purified recombinant fragment of human AFAP1L2 (AA: 674-818) expressed in E. Coli. | |
Primary | |
89.8 kDa |
2H6D1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
Monoclonal | |
Purified | |
actin filament associated protein 1-like 2, actin filament-associated protein 1-like 2, AFAP1-like protein 2, FLJ14564, KIAA1914Em:AC005383.4, XB130CTB-1144G6.4 | |
AFAP1L2 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title