Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFAP1L2 Mouse anti-Human, Clone:4E12D7, Abnova™

Catalog No. 89333926 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-926 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-926 Supplier Abnova Corporation Supplier No. MAB17497
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human AFAP1L2.

Specifications

Antigen AFAP1L2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4E12D7
Conjugate Unconjugated
Description actin filament associated protein 1-like 2
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene AFAP1L2
Gene Alias FLJ14564, KIAA1914, XB130
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 674-818 of human AFAP1L2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 84632
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.