Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFDN-DT Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen AFDN-DT
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123345
SDP
View Documents
Novus Biologicals
NBP309222100UL
100 μg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

AFDN-DT Polyclonal specifically detects AFDN-DT in Human samples. It is validated for Western Blot.
Specifications

Specifications

AFDN-DT
Western Blot
Unconjugated
Rabbit
Human
653483
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
The immunogen is a synthetic peptide directed towards the C-terminal region of human MLLT4-AS1. Peptide sequence GGTGSGRAVLGLDGRCSIQMGAAGSGRAVLGSVRTGGAGSKWAVLGSAGD
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.