Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AFF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567082 Supplier Invitrogen™ Supplier No. PA567082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDLIKFIMSLK SFSDATAPTQ EKIFAVLCMR CQSILNMAMF RCKKDIAIKY SRTLNKHFES SSK Highest antigen sequence identity to the following orthologs - mouse 73%, rat 80%.

Proto-oncogene AF-4 (or FEL) is a product of a chromosomal aberration of the human gene AFF1, which is associated with acute leukemias. The fusion of AF-4 on chromosome band 4q21 with the mixed lineage leukemia (MLL or HRX) gene on 11q23 results in a MLL-AF-4 chimeric transcription factor in which AF-4 contributes transcriptional effector properties and requires cell-specific accessory factors. MLL is involved in several chromosomal translocations associated with acute myeloid and lymphoid leukemia. The MLL-AF-4 fusion protein is expressed in all normal hematopoietic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFF1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AFF1
Gene Accession No. P51825
Gene Alias 9630032B01Rik; AF4; AF4/FMR2 family member 1; AF4/FMR2 family, member 1; AFF1; ALL1-fused gene from chromosome 4 protein; AW319193; FEL; homolog of human MLLT2 unidentified; MLLT2; Mllt2h; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 2; myeloid/lymphoid or mixed-lineage leukemia translocated to 2 homolog; PBM1; pre-B-cell monocytic leukemia partner 1; protein AF-4; Protein FEL; Proto-oncogene AF4; Rob; robotic; translocated to, 2
Gene Symbols AFF1
Host Species Rabbit
Immunogen Recombinant Human AFF1. Recombinant protein control fragment (Product #RP-107722).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4299
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.