Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AFF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551834 Supplier Invitrogen™ Supplier No. PA551834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSTSDSNTDQ EETLQIKVLP PCIISGGNTA KSKEICGASL TLSTLMSSSG SNNNLSISNE EPTFSPIPVM QTEILSPLRD HENLKNLWVK IDLDLLSRVP GHSSLHAAPA KPDHKETATK PKRQTAVTAV EKPAPKGKRK H Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 76%.

AFF2 belongs to the AF4 family. Defects in AFF2 are the cause of FRAXE. The exact function of AFF2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AFF2
Gene Accession No. P51816
Gene Alias AF4/FMR2 family member 2; AF4/FMR2 family member 2; LOW QUALITY PROTEIN: AF4/FMR2 family member 2; AF4/FMR2 family, member 2; AFF2; Fmr2; FMR2P; fragile X E mental retardation syndrome protein; fragile X mental retardation 2; fragile X mental retardation 2 homolog; fragile X mental retardation 2 protein; Fragile X mental retardation protein 2 homolog; FRAXE; MRX2; OX19; Ox19 homolog; Oxh; Protein FMR-2; Protein Ox19; RGD1565592; si:ch211-76h4.1
Gene Symbols AFF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AFF2. Recombinant protein control fragment (Product #RP-92219).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2334
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.