Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AFF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540852 Supplier Invitrogen™ Supplier No. PA540852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MDLFDFFRDW DLEQQCHYEQ DRSALKKREW ERRNQEVQQE DDLFSSGFDL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.

AFF2 belongs to the AF4 family. Defects in AFF2 are the cause of FRAXE. The exact function of AFF2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFF2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AFF2
Gene Accession No. P51816
Gene Alias AF4/FMR2 family member 2; AF4/FMR2 family member 2; LOW QUALITY PROTEIN: AF4/FMR2 family member 2; AF4/FMR2 family, member 2; AFF2; Fmr2; FMR2P; fragile X E mental retardation syndrome protein; fragile X mental retardation 2; fragile X mental retardation 2 homolog; fragile X mental retardation 2 protein; Fragile X mental retardation protein 2 homolog; FRAXE; MRX2; OX19; Ox19 homolog; Oxh; Protein FMR-2; Protein Ox19; RGD1565592; si:ch211-76h4.1
Gene Symbols AFF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human AFF2 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2334
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.