Learn More
Invitrogen™ AFF4 Polyclonal Antibody

Description
Immunogen sequence: LGNYDEMKDF IGDRSIPKLV AIPKPTVPPS ADEKSNPNFF EQRHGGSHQS SKWTPVGPAP STSQSQKRSS GLQSGHSSQR TSAGSSSGTN SSGQRHDRES YNNSGSSSRK KGQHGSEHSK SRSSSPGKPQ AVSSLNSSHS RSHG Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 88%.
Specifications
Specifications
| Antigen | AFF4 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AFF4 |
| Gene Accession No. | Q9UHB7 |
| Gene Alias | AF4/FMR2 family member 4; AF4/FMR2 family, member 4; AF5Q31; Aff4; Alf4; ALL1 fused gene from 5q31; ALL1-fused gene from chromosome 5q31 protein; CHOPS; HSPC092; Laf4l; LOW QUALITY PROTEIN: AF4/FMR2 family member 4; major CDK9 elongation factor-associated protein; MCEF; Protein AF-5q31; RA_m002_jsmFBA6Br |
| Gene Symbols | AFF4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.