Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFF4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988097
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8097 Supplier Bioss Supplier No. BS7886R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the AF4 family of transcription factors involved in leukemia. It is a component of the positive transcription elongation factor b (P-TEFb) complex. A chromosomal translocation involving this gene and MLL gene on chromosome 11 is found in infant acute lymphoblastic leukemia with ins.

Specifications

Antigen AFF4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AFF4
Gene Accession No. Q9UHB7
Gene Alias AF4/FMR2 family member 4; AF4/FMR2 family, member 4; AF5Q31; Aff4; Alf4; ALL1 fused gene from 5q31; ALL1-fused gene from chromosome 5q31 protein; BOS_8082; CHOPS; HSPC092; Laf4l; LOW QUALITY PROTEIN: AF4/FMR2 family member 4; Major CDK9 elongation factor-associated protein; MCEF; Protein AF-5q31; RA_m002_jsmFBA6Br
Gene Symbols AFF4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AFF4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27125
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.