Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AFG3L2 Antibody (OTI4B6), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271647G
Description
AFG3L2 Monoclonal specifically detects AFG3L2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
AFG3L2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AFG3L2 | |
Recombinant protein fragment corresponding to amino acids 1-250 of human AFG3L2 (NP_006787) produced in E.coli. | |
0.1 mL | |
10939.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4B6 | |
Western Blot | |
AFG3 (ATPase family gene 3, yeast)-like 2, AFG3 ATPase family gene 3-like 2 (S. cerevisiae), AFG3 ATPase family gene 3-like 2 (yeast), AFG3-like protein 2, ATPase family gene 3, yeast, EC 3.4.24, EC 3.4.24.-, FLJ25993, Paraplegin-like protein, SCA28, spinocerebellar ataxia 28 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction