Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AFG3L2 Antibody (OTI4B6), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271647C
Description
AFG3L2 Monoclonal specifically detects AFG3L2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| AFG3L2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AFG3L2 | |
| Recombinant protein fragment corresponding to amino acids 1-250 of human AFG3L2 (NP_006787) produced in E.coli. | |
| 0.1 mL | |
| 10939.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI4B6 | |
| Western Blot | |
| AFG3 (ATPase family gene 3, yeast)-like 2, AFG3 ATPase family gene 3-like 2 (S. cerevisiae), AFG3 ATPase family gene 3-like 2 (yeast), AFG3-like protein 2, ATPase family gene 3, yeast, EC 3.4.24, EC 3.4.24.-, FLJ25993, Paraplegin-like protein, SCA28, spinocerebellar ataxia 28 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction