Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AFG3L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590436 Supplier Invitrogen™ Supplier No. PA590436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSINQKHFEQ AIERVIGGLE KKTQVLQPEE KKTVAYHEAG HAVAGWYLEH ADPLLKVSII PRGKGLGYAQ YLPKEQYLYT KEQLLDRMCM TLGGRVSEEI FFGRITTGAQ DDLRKVTQSA YAQIVQFGMN EKVGQISFDL PRQGDMVLEK PYSEATARLI DDEVRILIND AYKRTVALLT EKKADVEKVA LLLLEKEVLD KNDMVELLGP RPFAEKSTYE EFVEGTGSLD EDTSLPEGLK DWNKEREKEK EEPPGEKVAN; Positive Samples: LO2, 293T, HeLa, Mouse brain, Mouse heart, Mouse kidney, Mouse liver, Mouse pancreas; Cellular Location: Mitochondrion membrane, Multi-pass membrane protein.

AFG3L2 is a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. AFG3L2 gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders. This gene encodes a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. This gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFG3L2
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene AFG3L2
Gene Accession No. Q8JZQ2, Q9Y4W6
Gene Alias 2310036I02Rik; AFG3 ATPase family gene 3-like 2; AFG3 ATPase family member 3-like 2; AFG3 like AAA ATPase 2; AFG3 like matrix AAA peptidase subunit 2; AFG3(ATPase family gene 3)-like 2; AFG3L2; AFG3-like AAA ATPase 2; AFG3-like protein 2; AFG3-like protein 2 (Paraplegin-like protein); ATPase family gene 3, yeast; AW260507; Emv66; FLJ25993; par; paraplegin-like protein; SCA28; si:ch211-12e1.4; similar to AFG3-like protein 2 (Paraplegin-like protein); SPAX5
Gene Symbols AFG3L2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 538-797 of human AFG3L2 (NP_0067872).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10939, 307350, 69597
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.