Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AFG3L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598275
Description
AFG3L2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
AFG3L2 is a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. AFG3L2 gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders. This gene encodes a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. This gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders.
Specifications
AFG3L2 | |
Polyclonal | |
Unconjugated | |
AFG3L2 | |
2310036I02Rik; AFG3 ATPase family gene 3-like 2; AFG3 ATPase family member 3-like 2; AFG3 like AAA ATPase 2; AFG3 like matrix AAA peptidase subunit 2; AFG3(ATPase family gene 3)-like 2; AFG3L2; AFG3-like AAA ATPase 2; AFG3-like protein 2; AFG3-like protein 2 (Paraplegin-like protein); ATPase family gene 3, yeast; AW260507; Emv66; FLJ25993; par; paraplegin-like protein; SCA28; si:ch211-12e1.4; similar to AFG3-like protein 2 (Paraplegin-like protein); SPAX5 | |
Rabbit | |
Protein G | |
RUO | |
10939 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9Y4W6 | |
AFG3L2 | |
Recombinant Human AFG3-like protein 2 protein (550-759AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction