Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Afinion™ HbA1c Control Rx

Catalog No. 23-050-106 Shop All Abbott Rapid Diagnostics Products
Click to view available options
Quantity:
(2) 0.5 mL/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Afinion™ HbA1c Control contains liquid preparations of Stabilized Porcine Whole Blood (Control CI) and Human Whole Blood (Control CII). Designed for use with the Afinion™ Analyzer, the Afinion™ HbA1c Test, and the Afinion™ HbA1c Dx Test.

  • Capillary and venous whole blood sample material
  • 1.5 μL sample volume
  • Measuring range: 4% to 15% HbA1c
  • Storage: 3 months at room temperature
  • Ready-to-use controls in two levels

Specifications

Certifications/Compliance NGSP and IFCC Certified
Compatible Methods Traceable to the IFCC Reference Method
Content And Storage Up to 3 months (at Room Temperature)
For Use With (Equipment) Afinion™ Analyzer, Afinion™ HbA1c Test, and Afinion™ HbA1c Dx Test
Type HbA1c Control
Quantity (2) 0.5 mL/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.