Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aflatoxin B1 Antibody (6A10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280563
Description
Aflatoxin B1 Monoclonal specifically detects Aflatoxin B1 in Human, All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.Specifications
Aflatoxin B1 | |
Monoclonal | |
1.5 mg/mL | |
Western Blot 1:500-1:2000, ELISA : 5 μg/mL, Immunohistochemistry : 1:100, Immunocytochemistry/Immunofluorescence : 1:100, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot | |
AFB1, AFB1-AR1, Aldoketoreductase 7 | |
Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1). | |
0.1 mL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
6A10 | |
Unconjugated | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Human, All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction