Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aflatoxin B1 Antibody (6A10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB600443H
Description
Aflatoxin B1 Monoclonal specifically detects Aflatoxin B1 in Human, All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.Specifications
| Aflatoxin B1 | |
| Monoclonal | |
| HRP | |
| AFB1, AFB1-AR1, Aldoketoreductase 7 | |
| Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1). | |
| 0.1 mL | |
| Cancer | |
| All species. There is no cross-reactivity with other carcinogen-DNA adducts. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot | |
| 6A10 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction