Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aflatoxin Mix 4 Solution, 0.5 μg/mL B 2 and G 2 in Acetonitrile, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
2 mL
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications
Specifications
CAS | 75-05-8 |
MDL Number | MFCD04974643 |
Type | Analytical Standard |
Quantity | 1 mL |
Grade | Analytical Standard |
Concentration or Composition (by Analyte or Components) | 0.5 μg/mL B 2 and G 2 in. acetonitrile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction