Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aflatoxin Monoclonal Antibody (6A10), Biotin

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516119
Description
This antibody does not cross-react with other carcinogen-DNA adducts. Suggested positive control: human HepG2 liver samples.
Aflatoxin is a naturally occurring mycotoxin produced by two types of mold: Aspergillus flavus and Aspergillus parasiticus. Aspergillus flavus is common and widespread in nature and is most often found when certain grains are grown under stressful conditions such as drought. The mold occurs in soil, decaying vegetation, hay, and grains undergoing microbiological deterioration and invades all types of organic substrates whenever and wherever the conditions are favorable for its growth. Favorable conditions include high moisture content and high temperature. At least 13 different types of aflatoxin are produced in nature with aflatoxin B1 considered as the most toxic. While the presence of Aspergillus flavus does not always indicate harmful levels of aflatoxin it does mean that the potential for aflatoxin production is present.
Specifications
| Aflatoxin | |
| Monoclonal | |
| 0.79 mg/mL | |
| PBS with 0.05% sodium azide | |
| Aflatoxin B1 DNA (against the imidazole ring-opened persistent form of the major N-7 guanine adduct of AFB1). | |
| 100 μL | |
| Primary | |
| 4°C, store in dark | |
| Liquid | |
| Chemical |
| Dot Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6A10 | |
| Biotin | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction