Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aflatoxin Monoclonal Antibody (6A10), HRP
GREENER_CHOICE

Catalog No. PIMA516122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516122 Supplier Invitrogen™ Supplier No. MA516122
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not cross-react with other carcinogen-DNA adducts. Suggested positive control: human HepG2 liver samples.

Aflatoxin is a naturally occurring mycotoxin produced by two types of mold: Aspergillus flavus and Aspergillus parasiticus. Aspergillus flavus is common and widespread in nature and is most often found when certain grains are grown under stressful conditions such as drought. The mold occurs in soil, decaying vegetation, hay, and grains undergoing microbiological deterioration and invades all types of organic substrates whenever and wherever the conditions are favorable for its growth. Favorable conditions include high moisture content and high temperature. At least 13 different types of aflatoxin are produced in nature with aflatoxin B1 considered as the most toxic. While the presence of Aspergillus flavus does not always indicate harmful levels of aflatoxin it does mean that the potential for aflatoxin production is present.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aflatoxin
Applications Dot Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6A10
Concentration 0.63 mg/mL
Conjugate HRP
Formulation PBS with no preservative
Host Species Mouse
Immunogen Aflatoxin B1 DNA (against the imidazole ring-opened persistent form of the major N-7 guanine adduct of AFB1).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.