Learn More
Invitrogen™ AFM Polyclonal Antibody

Description
Immunogen sequence: EDVSSNYDGC CEGDVVQCIR DTSKVMNHIC SKQDSISSKI KECCEKKIPE RGQCIINSNK DDRPKDLSLR EGKFTDSENV CQERDADPDT FFAKFTFEYS RRHPDLSIPE LLRIVQIYKD LLRNCCNTEN PPGCYRYAED KFNETTEKSL KMVQQECKHF QNLGKDGLKY HYLIRLTKIA PQLSTEELVS LGEKMVTAFT TCCTLSEEFA CVDNLADLVF GELCGVNENR TINPAVDHCC KTNFAFRRPC FESLKADKTY VPPPFSQDLF TFHADMCQSQ NEELQRKTDR FLVNLVKLKH ELTDEELQSL FTNFANVVDK CCKAESPEVC FNEESPKIGN.
Specifications
Specifications
| Antigen | AFM |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.07 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | AFM |
| Gene Accession No. | P43652 |
| Gene Alias | Afamin; AFM; ALB2; ALBA; Alf; alpha albumin; alpha-Alb; Alpha-albumin; MGC125338; MGC125339 |
| Gene Symbols | AFM |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.