Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AFMID Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100379 Supplier Invitrogen™ Supplier No. PA5100379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AFMID.

Catalyzes the hydrolysis of N-formyl-L-kynurenine to L-kynurenine, the second step in the kynurenine pathway of tryptophan degradation. Kynurenine may be further oxidized to nicotinic acid, NAD(H) and NADP(H). Required for elimination of toxic metabolites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFMID
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AFMID
Gene Accession No. Q63HM1, Q8K4H1
Gene Alias 9030621K19Rik; AFMID; Ammd; Arylformamidase; FKF; formylase; formylkynureninase; hypothetical protein LOC550372; im:7148587; Kf; KFA; KFase; kynurenine formamidase; N-formylkynurenine formamidase; probable arylformamidase; zgc:112472
Gene Symbols AFMID
Host Species Rabbit
Immunogen A synthesized peptide derived from human AFMID(Accession Q63HM1), corresponding to amino acid residues T142-L192.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125061, 71562
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.