Learn More
Invitrogen™ AFP Monoclonal Antibody (C2)

Description
C2 has been characterized in the ISOBM TD-2 workshop and assigned by K. Nustad to group E of a cluster of 6 major epitopes of human alpha fetoprotein. Human alpha fetoprotein is an oncofetal protein of 70 kDa. It is expressed in fetal liver and is normally absent in health adult tissues. It is positive on all yolk sac tumors, on some other germ cell tumors and on hepatocellular carcinomas.
Specifications
Specifications
| Antigen | AFP |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | C2 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | AFP |
| Gene Accession No. | P02771, P02772 |
| Gene Alias | a fetoglobulin; a fetoprotein; Afp; AFPD; Alpha fetoglobulin; alpha fetoprotein; alpha-1-fetoprotein; Alpha-fetoglobulin; alpha-fetoprotein; alpha-fetoprotein precursor; alpha-foetoprotein; chloride intracellular channel 6; FETA; fetoglobulin; fetoprotein; HPAFP; a fetoglobulin; a fetoprotein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.