Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFP Mouse anti-Human, Unconjugated, Clone: C3, Abnova™

Catalog No. 89337404 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-404 Supplier Abnova Corporation Supplier No. MAB20909
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

AFP Monoclonal antibody specifically detects AFP in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen AFP
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone C3
Conjugate Unconjugated
Description alpha-fetoprotein
Gene AFP
Gene Alias FETA, HPAFP
Host Species Mouse
Immunogen Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 174
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.