Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFP Polyclonal Antibody, Huabio

Rabbit Polyclonal Antibody

Supplier:  HUABIO RESEARCH INC R16011100UL

Encompass
Encompass_Preferred

Catalog No. 50-254-4378


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

AFP Polyclonal antibody specifically detects AFP in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

AFP
Polyclonal
Unconjugated
AFP
a fetoglobulin; a fetoprotein; Afp; AFPD; Alpha fetoglobulin; alpha fetoprotein; alpha-1-fetoprotein; Alpha-fetoglobulin; alpha-fetoprotein; alpha-fetoprotein precursor; alpha-foetoprotein; chloride intracellular channel 6; FETA; fetoglobulin; fetoprotein; HPAFP; a fetoglobulin; a fetoprotein
Recombinant protein within N-terminal human AFP.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
P02771, P02772, P02773
Rabbit
Protein A
RUO
11576, 174, 24177
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.