Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
African Swine Fever Virus p54 Antibody (HL1289) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325370
Description
African Swine Fever Virus p54 Monoclonal antibody specifically detects African Swine Fever Virus p54 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA, Lateral FlowSpecifications
African Swine Fever Virus p54 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
22220355 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunocytochemistry, ELISA, Lateral Flow | |
HL1289 | |
Western Blot 1:5000-1:100000, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA, Lateral Flow Assay | |
ASFV p54, Ba71V-126, E183L, Envelope protein p54, Structural protein p54 | |
Recombinant protein encompassing a sequence within the center region of African Swine Fever Virus p54. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction