Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ African Swine Fever Virus Structural Protein p54 Monoclonal Antibody (2G6G10)
GREENER_CHOICE

Catalog No. FEMA550287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550287 Supplier Invitrogen™ Supplier No. MA550287
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

African Swine Fever Virus Structural Protein p54 Monoclonal Antibody for ELISA

The African Swine Fever Virus (ASFV) structural protein p54, encoded by the E183L gene, is a crucial component of the virus's architecture, playing a significant role in its morphogenesis. This 25-kDa polypeptide is essential for the recruitment of envelope precursors to assembly sites, contributing to the formation of viral particles. It is highly antigenic and has been localized within the replication factories of infected cells, predominantly associated with membranes and immature virus particles. The protein p54 has been shown to interact with target cells, aiding in the virus's attachment and entry, alongside other viral proteins like p30. Attempts to inactivate the p54 gene have demonstrated its necessity for ASFV replication, underscoring its potential as a target for therapeutic interventions. Its structural and functional properties make p54 a focal point of study in the development of ASFV treatments and prevention strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen African Swine Fever Virus Structural Protein p54
Applications ELISA, Neutralization
Classification Monoclonal
Clone 2G6G10
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Ba71V-126
Gene Accession No. Q65194
Gene Alias African swine fever virus p54
Gene Symbols Ba71V-126, E183L
Host Species Mouse
Immunogen Recombinant Envelope protein p54 protein (54-183AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22220355
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.