Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ African Swine Fever Virus Structural Protein p54 Recombinant Rabbit Monoclonal Antibody (HL1286)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA546989
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The African Swine Fever Virus (ASFV) structural protein p54, encoded by the E183L gene, is a crucial component of the virus's architecture, playing a significant role in its morphogenesis. This 25-kDa polypeptide is essential for the recruitment of envelope precursors to assembly sites, contributing to the formation of viral particles. It is highly antigenic and has been localized within the replication factories of infected cells, predominantly associated with membranes and immature virus particles. The protein p54 has been shown to interact with target cells, aiding in the virus's attachment and entry, alongside other viral proteins like p30. Attempts to inactivate the p54 gene have demonstrated its necessity for ASFV replication, underscoring its potential as a target for therapeutic interventions. Its structural and functional properties make p54 a focal point of study in the development of ASFV treatments and prevention strategies.
Specifications
African Swine Fever Virus Structural Protein p54 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q65194 | |
Ba71V-126, E183L | |
Recombinant protein encompassing a sequence within the center region of ASFV p54. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Virus | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
HL1286 | |
Unconjugated | |
Ba71V-126 | |
African swine fever virus p54 | |
Rabbit | |
Protein A | |
RUO | |
22220355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction