Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ African Swine Fever Virus Structural Protein p54 Recombinant Rabbit Monoclonal Antibody (HL1218)
GREENER_CHOICE

Catalog No. FEMA550304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550304 Supplier Invitrogen™ Supplier No. MA550304
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product can be used in Sandwich ELISA to detect African Swine Fever Virus Structural Protein p54 using MA5-50304 as a capture antibody and MA5-50305 as the detection antibody.

The African Swine Fever Virus (ASFV) structural protein p54, encoded by the E183L gene, is a crucial component of the virus's architecture, playing a significant role in its morphogenesis. This 25-kDa polypeptide is essential for the recruitment of envelope precursors to assembly sites, contributing to the formation of viral particles. It is highly antigenic and has been localized within the replication factories of infected cells, predominantly associated with membranes and immature virus particles. The protein p54 has been shown to interact with target cells, aiding in the virus's attachment and entry, alongside other viral proteins like p30. Attempts to inactivate the p54 gene have demonstrated its necessity for ASFV replication, underscoring its potential as a target for therapeutic interventions. Its structural and functional properties make p54 a focal point of study in the development of ASFV treatments and prevention strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen African Swine Fever Virus Structural Protein p54
Applications ELISA, Lateral Flow, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1218
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ba71V-126
Gene Accession No. Q65194
Gene Alias African swine fever virus p54
Gene Symbols Ba71V-126, E183L
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of ASFV p54. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22220355
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.