Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    AG-2/AGR2 Antibody (10E2), Alexa Fluor™ 750, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227175AF750
Description
AG-2/AGR2 Monoclonal specifically detects AG-2/AGR2 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AG-2/AGR2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| AGR2 | |
| A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Bovine, Canine, Equine, Primate | |
| IgG1 κ | 
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1000 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
| Mouse | |
| Protein G purified | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 10551.0 | |
| Store at 4°C in the dark. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            