Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG-2/AGR2 Antibody (10E2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227175FR
Description
AG-2/AGR2 Monoclonal specifically detects AG-2/AGR2 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AG-2/AGR2 | |
Monoclonal | |
DyLight 680 | |
O95994 | |
AGR2 | |
A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody. | |
0.1 mL | |
Breast Cancer, Cancer, Stem Cell Markers | |
10551 | |
Human, Mouse, Bovine, Canine, Equine, Other, Primate | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1000 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Primate/Monkey, Canine/Dog, Mouse, Bovine, Horse, Orangutan, Panda and Platypus. Immunogen sequence similarity with other species: Xenopus (82%), Chicken (83%). | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction